Learn More
Invitrogen™ TrackIt™ Cyan/Orange Loading Buffer
A 6X solution used for easy loading and tracking of DNA samples in agarose gels
Brand: Invitrogen™ 10482028
28.48 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00454kg
Includes: TrackIt™ Cyan/Orange Loading Buffer is composed of 30% (v/v) glycerol, 60mM Tris-HCl (pH 7.5), 60mM EDTA, 0.36% (w/v) XCFF, and 2.4% (w/v) Orange G. TrackIt™ Cyan/Yellow Loading Buffer is composed of 30% (v/v) glycerol, 60mM Tris-HCl (pH 7.5), 60mM EDTA, 0.36% (w/v) XCFF, and 3.6% (w/v) Tartrazine
Description
TrackIt™ Loading Buffers loading buffers contain two tracking dyes, one that runs behind sample and one that runs ahead of sample. The tracking dyes serve as visual markers for following migration during electrophoresis and indicate when maximum resolution of DNA fragments has been achieved. Additionally, visualization of DNA bands will not be obscured by tracking dyes because they run outside limits of most DNA samples.
- TrackIt Cyan/Orange Loading Buffer is recommended for DNA fragments between 50 bp and 10 Kb
- TrackIt Cyan/Yellow Loading Buffer is recommended for DNA fragments between 10 bp and 1 Kb
- For use with agarose and E-Gel™ gels
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Approved for shipment at Room Temperature or on Wet or Dry Ice
Specifications
• 3 x 0.5 mL 6X TrackIt Cyan/Orange Loading Buffer Store at room temperature or at 4°C for up to 6 months. For longer periods, store at -20°C. |
|
6X | |
Orange G, Xylene Cyanol, Xylene Cyanol FF (XCFF) | |
0.5 mL | |
TrackIt™ |
Sample Loading Buffers | |
Agarose | |
Loading Buffer | |
Agarose Gels | |
Approved for shipment at Room Temperature or on Wet or Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.