Learn More
Invitrogen™ UltraPure™ 0.5M EDTA, pH 8.0
Sterile-filtered solution
Brand: Invitrogen™ 15575020
48.00 GBP valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Includes: 4 x 100mL bottles UltraPure 0.5M EDTA, pH 8.0.
Description
- Aqueous solution prepared by dissolving Na2EDTA•2H2O in distilled, deionized water and adjusting the pH to 8.0 with sodium hydroxide
- Suitable for biochemistry or molecular biology applications requiring chelator of divalent metal ions
Agarose Gel Electrophoresis, ChIP-on-Chip, Chromatin Biology, DNA & RNA Purification & Analysis, DNA Extraction, General gDNA Purification Reagents & Accessories, Genomic DNA Purification, Maxiprep, Midiprep, Miniprep, Nucleic Acid Gel Electrophoresis & Blotting, Plasmid DNA Purification, RNAi, Epigenetics & Non-Coding RNA Research
Order Info
Shipping Condition: Room Temperature

Specifications
EDTA | |
Contents: 4 bottles (100 mL each) of UltraPure™ 0.5M EDTA, pH 8.0. Store at room temperature. | |
pH 8.0 | |
EDTA | |
Molecular Biology Grade | |
Liquid |
0.5 M | |
Chromatin Biology, Nucleic Acid Gel Electrophoresis, Blotting | |
UltraPure™ | |
4 x 100 mL | |
Room Temperature |
Safety and Handling
- UltraPure 0.5M EDTA
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P314-Get medical advice/attention if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium hydroxide, disodium EDTA
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.