missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ 1 M Tris-HCI Buffer, pH 7.5
GREENER_CHOICE

Product Code. 10123722
Click to view available options
Quantity:
1 L
Unit Size:
1 L
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10123722

Brand: Invitrogen™ 15567027

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

UltraPure 1M Tris-HCl, pH 7.5

Pre-mixed, pH-adjusted, sterile-filtered solution

  • Prepared as 1M concentrate, buffer can be diluted to desired concentration
  • Used in molecular biology or general biochemistry applications
  • Performance and quality testing: No DNase, RNase, or protease activity detected
  • Sustainable packaging

Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, DNA Extraction, General gDNA Purification Reagents & Accessories, Genomic DNA Purification, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 M
Content And Storage Store in refrigerator (2-8°C). UltraPure™ 1M Tris-HCl, pH 7.5 is guaranteed for 6 months from date of purchase unless otherwise indicated in product documentation.
pH pH 7.5
Product Line UltraPure
Product Type Tris-HCl Buffer
Quantity 1 L
Shipping Condition Room Temperature
Form Liquid
compliance-icons
Product Identifier
  • Di-2-pyridyl ketone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Bis(2-pyridyl) ketone

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.