missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Ethidium Bromide, 10 mg/mL

Product Code. 11508756
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11508756

Brand: Invitrogen™ 15585011

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Includes

10mL of 10mg/mL solution

Sensitive fluorescent dye detects nucleic acids in agarose gels and cesium chloride gradients

UltraPure Ethidium Bromide is available as an aqueous solution containing 10mg/mL UltraPure ethidium bromide and high-purity water. Sensitive: Detects as little as little as 1ng of nucleic acids in agarose gels.

  • Versatile: Detects DNA or RNA
  • Easy to Use: Can be used undiluted or diluted to the desired concentration
  • An aromatic compound with a phenanthridine core, ethidium bromide has excitation maxima at 300nm and 520nm and an emission maximum at 600nm

Agarose Gel Electrophoresis, DNA & RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 10ml of 10mg⁄ml solution. Store at 15°C to 30°C
Detection Location In-Gel Detection
Detection Method Fluorescence
Label or Dye Ethidium bromide
Product Type Ethidium Bromide
Product Line UltraPure
Quantity 10 mL
Shipping Condition Room Temperature
Target Molecule RNA, DNA
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.