Learn More
Invitrogen™ UltraPure™ DNA Typing Grade™ 50X TAE Buffer
Most commonly used buffer for agarose DNA electrophoresis
Brand: Invitrogen™ 24710030
166.56 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : Weight : 1.00000kg
Description
UltraPure DNA Typing Grade™ 50X TAE Buffer is a sterile-filtered solution of 2M Tris-acetate and 50 mM EDTA. To prepare a 1X solution, mix one volume of UltraPure 50X TAE Buffer with 49 volumes of distilled water.
- Contains 40mM Tris-acetate and 1mM EDTA with a pH of 8.3 at 23°C.
- Performance and Quality Testing: No DNase activity detected
- Compatible with agarose and E-Gel™ gels
- Sustainable packaging
Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Running Buffer | |
Store at room temperature. | |
Nucleic Acid Gel Electrophoresis, Blotting | |
UltraPure™ | |
1 L |
50 X | |
E-Gel™, Agarose Gels | |
Sustainable packaging | |
TAE Buffer | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.