Learn More
Invitrogen™ UltraPure™ Glycerol
Description
UltraPure Glycerol (≥99% pure) is suitable for concentrating and storing enzymes at low temperatures. It can be diluted to a 50% (v/v) solution that will not freeze at -20°C. With a density of 1.26 g/mL, it is commonly used in electrophoresis loading buffers. Additionally, UltraPure Glycerol gradients are ideal for purifying proteins, bacteriophage, and organelles.
Performance and quality testing: no DNase, RNase, or protease activity detected
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Chemical Name or Material | Glycerol |
| Content And Storage | Store at room temperature. |
| Product Line | UltraPure |
| Purity | Molecular Biology Grade |
| Quantity | 500 mL |
| Shipping Condition | Room Temperature |
| Form | Liquid |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.