missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Glycogen

Product Code. 10757844
Click to view available options
Quantity:
100 μL
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10757844

Brand: Invitrogen™ 10814010

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

1 vial containing 100μL of 20μg/μL ultrapure glycogen

Used as carrier for nucleic acid precipitation in ethanol

  • Concentration: 20μg/μL in RNase-free water
  • Performance and quality testing: No DNase or RNase activity detected

ChIP-on-Chip, Chromatin Biology, DNA & RNA Purification & Analysis, DNA Extraction, General gDNA Purification Reagents & Accessories, Genomic DNA Purification, RNAi, Epigenetics & Noncoding RNA Research

Order Info

Shipping Conditions: Approved for shipment on wet or dry ice

TRUSTED_SUSTAINABILITY

Specifications

Grade Molecular Biology
Content And Storage 1 vial containing 100 μl of 20 μg/μl ultrapure glycogen.

Store at -5°C to -30°C.
Shipping Condition Approved for shipment on Wet or Dry Ice
Quantity 100 μL
Product Type Glycogen
Product Line UltraPure
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.