missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UltraPure™ Phenol:Chloroform:Isoamyl Alcohol (25:24:1, v/v)
UltraPure™ Phenol:Chloroform:Isoamyl Alcohol (25:24:1, v/v) is used in the purification of nucleic acids. This reagent consists of highly pure chloroform, isoamyl alcohol, and UltraPure™ Phenol saturated with Tris-HCl.
Brand: Invitrogen™ 15593031
38.15 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
View more versions of this product
This item is not returnable.
View return policy
Description
- Reagent consists of highly pure chloroform, isoamyl alcohol, and UltraPure™ Phenol saturated with Tris-HCl
- When mixtures are extracted with Phenol:Chloroform:Isoamyl Alcohol, proteins are denatured and collected in the organic phase or at the interphase, while nucleic acids remain in the aqueous phase
- UltraPure™ Phenol:Chloroform:lsoamyl Alcohol contains no preservatives
- It is packaged under an inert gas in shatter-resistant, plastic-coated amber bottles
- The appearance of the solution is evaluated at room temperature
- No RNase or DNase activity is detected
- Store in the refrigerator at 2°C to 8°C
Order Info
Shipping Condition: Room temperature
Specifications
Phenols | |
8.05 | |
Molecular Biology Grade | |
Room Temperature |
Store at 2°C to 8°C. To extend product stability, it is recommended that the gas volume above the solution be flushed with argon or nitrogen following each use. | |
UltraPure™ | |
100 mL | |
Liquid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- UltraPure Phenol:Chloroform:Isoamyl Alcohol (25:24:1, v/v)
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after repeated exposure Category 2
- Carcinogenicity Category 2
- Germ cell mutagenicity Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H373-May cause damage to organs.
- H341-Suspected of causing genetic defects.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P264-Wash thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Phenol, Chloroform,1-Butanol, 3-methyl-
For Research Use Only. Not for use in diagnostic procedures.