missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Salmon Sperm DNA Solution

Product Code. 10605543
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
Pack of 5
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10605543

Brand: Invitrogen™ 15632011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

5 tubes Salmon Sperm DNA solution (1mL each, 10mg/mL)

Used directly in preparation of prehybridization and hybridization solutions

UltraPure™ Salmon Sperm DNA Solution is a ready-to-use, sheared DNA solution that is used directly in the preparation of prehybridization and hybridization solutions.

  • Prepared from highly pure, phenol/chloroform-extracted DNA and DNase-free, RNase-free (DEPC-treated), distilled, deionized water
  • DNA is sheared to average size of ≤2,000 bp
  • Concentration is adjusted to 10 mg/mL
  • Used to block nonspecific attachment of probe to surface of membrane

ChIP-on-Chip, Chromatin Biology, DNA & RNA Purification & Analysis, Gene Expression Analysis & Genotyping, Microarray Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis & Blotting, RNAi, Epigenetics & Non-Coding RNA Research, Southern Blotting

Order Info

Shipping Conditions: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 5 tubes Salmon Sperm DNA solution (1 ml each, 10 mg/ml)

UltraPure™ Salmon Sperm DNA is stable for 1 year at -20°C.
For Use With (Application) Chromatin Biology, Nucleic Acid Gel Electrophoresis, Blotting
Key Functions Block Non-Specific Hybridization
Product Line UltraPure
Product Type Salmon Sperm DNA Solution
Quantity 5 x 1 mL
Reagent Type Salmon Sperm DNA
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.