Learn More
Invitrogen™ UltraPure™ SDS Solution, 10%
Filtered and ready to use
£83.75 - £160.00
Specifications
Product Type | SDS Solution |
---|---|
Form | Liquid |
Green Features | Sustainable packaging |
Product Line | UltraPure™ |
Reagent Type | SDS |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11538896
|
Invitrogen™
15553027 |
4 x 100 mL |
£83.75
400mL |
In Stock
Log in to see stock. |
|||||
10154042
|
Invitrogen™
24730020 |
1 L |
£160.00
1 L |
In Stock
Log in to see stock. |
|||||
Description
UltraPure 10% SDS Solution is a premixed aqueous solution containing 10% sodium dodecyl sulfate (SDS). SDS is a protein denaturant used in polyacrylamide gel electrophoresis for molecular weight determinations.
- Used to dissociate nucleic acid-protein complexes in DNA extraction protocols, to disrupt cell membranes, and to prepare prehybridization and hybridization solutions
- Sustainable packaging
DNA & RNA Purification & Analysis, DNA Extraction, Gene Expression Analysis & Genotyping, General gDNA Purification Reagents & Accessories, Genomic DNA Purification, Microarray Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis & Blotting, Southern Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
SDS Solution | |
Sustainable packaging | |
SDS |
Liquid | |
UltraPure™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.