missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Sucrose

UltraPure Sucrose is suitable for forming density gradients for a variety of separation applications.

Brand:  Invitrogen™ 15503022

130.90 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 11538736

  • £187.00 / 5kg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

UltraPure Sucrose is suitable for forming density gradients for a variety of separation applications. It is used to separate RNAs, to prepare bacteriophage λ-DNA arms, and to separate proteins. UltraPure Sucrose gradients typically are formed by use of gradient formers or by layering two or more sucrose solutions in varying amounts in a centrifuge tube.

Performance and Quality Testing
No protease activity detected.

Order Info

Approved for shipment at Room Temperature or on Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sucrose
Density Gradients - Separation Applications
Molecular Biology Grade
Approved for shipment at Room Temperature or on Wet Ice
Store at room temperature.
UltraPure™
5 kg
Powder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.