missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ TAE Buffer, 10X
GREENER_CHOICE

Product Code. p-4926452
Click to view available options
Quantity:
1 L
4 L
Unit Size:
1 L
4 L
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10154672

Brand: Invitrogen™ 15558042

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Compatible with agarose gels

  • UltraPure 10X TAE Buffer is sterile-filtered solution of 400 mM Tris-acetate and 10 mM EDTA
  • TAE Buffer is most commonly used buffer for agarose DNA electrophoresis
  • Supplied in 1 L plastic bottles/4 L/10 L stackable Cubitainer™ Box
  • 1X TAE Buffer solution contains 40 mM Tris-acetate and 1mM EDTA at pH 8.3
  • Performance and Quality Testing: No DNase activity detected. Sustainable packaging

Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Buffer Running Buffer
Concentration 10 X
Content And Storage Store at room temperature.
Format Bottle
Gel Compatibility Agarose Gels
For Use With (Application) Nucleic Acid Gel Electrophoresis, Blotting
Product Line UltraPure
Product Type TAE Buffer
Quantity 1 L
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.