missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Tris Buffer (powder format)

Product Code. 10533243
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10533243

Brand: Invitrogen™ 15504020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

One 1kg container UltraPure Tris buffering agent

No DNase, RNase, or protease activity detected

  • Used because of its buffering range (pH 7.0 to 9.0) and compatibility with many enzymes, including restriction endonucleases and DNA-modifying enzymes

Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Condition: Approved for shipment at room temperature or on wet ice

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris
Content And Storage Contents: One container (1 kg) of UltraPure™ Tris buffering agent
For Use With (Application) Nucleic Acid Gel Electrophoresis, Blotting
Product Line UltraPure
Product Type Tris Buffer
Quantity 1 kg
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Form Powder

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.