missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Agarose-1000
GREENER_CHOICE

Product Code. 10460755
Click to view available options
Quantity:
100 g
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10460755

Brand: Invitrogen™ 16550100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

100g pouch

Easy-pour spout reduces spills and contamination

UltraPure Agarose-1000 is polysaccharide used for size-based separation of nucleic acids in agarose gel electrophoresis. UltraPure Agarose 1000 is specifically formulated for the high-resolution separation of small (<1,000 bp) DNA, RNA, and PCR fragments. This standard melting temperature agarose can resolve DNA fragments differing in as little as 10bp in size

  • Ideal for analysis and recovery of small DNA and RNA fragments
  • Strong gel structure allows for better handling
  • Improved clarity allows for enhanced visibility of fragments
  • Improved packaging: Environmentally friendly, sustainable packaging uses 75% less plastic than the original bottles
  • Performance and quality testing: Performance of UltraPure Agarose 1000 is evaluated to satisfy specifications in appearance, moisture, gel strength, gelling temperature, melting temperature, sulfate content, electroendosmosis, and DNase/RNase assay Separation range: 50bp to 1000bp

UltraPure Agarose 10000 (Cat. No. 16550-100) is a replacement for Cat. No. 10975-035.

Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 pouch containing 100 g of UltraPure™ Low Melting Point Agarose. Store at room temperature.
Product Type Agarose
Separation Range 50 to 1000 bp
Form Powder
For Use With (Application) Nucleic Acid Gel Electrophoresis, Blotting
Gel Compatibility Agarose Gels
Green Features Sustainable packaging
Melting Point Standard Melting Point
Product Line UltraPure
Quantity 100 g
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.