missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Agarose
GREENER_CHOICE

Product Code. 10264544
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10264544

Brand: Invitrogen™ 16500500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Easy-pour spout reduces spills and contamination

UltraPure™ Agarose is a polysaccharide used for size-based separation of nucleic acids in agarose gel electrophoresis applications. UltraPure™ Agarose is ideal for resolving DNA and RNA fragments from 100 bp to >30 kb

  • Ideal for analysis and recovery of DNA and RNA for routine applications
  • Strong gel structure allows for better handling and less breakage
  • Can be used in protein electrophoresis applications such as Ouchterlony (antigen-antibody interaction assay) and radial immunodiffusion (RID) (antigen quantitation assay)
  • Improved packaging
  • Performance and quality testing

Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contents: 1 pouch containing 500 g of UltraPure™ Agarose
Storage: 15°C to 30°C
Product Type Agarose
Separation Range 100 to >30000 bp
Form Powder
For Use With (Application) Nucleic Acid Gel Electrophoresis, Protein Electrophoresis, Blotting
Gel Compatibility Agarose Gels
Green Features Sustainable packaging
Melting Point Standard Melting Point
No. of Reactions 2500
Product Line UltraPure
Quantity 500 g
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.