missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 293FT Cell Line

Product Code. 10769564
Click to view available options
Quantity:
1 x 107 cells
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10769564

Brand: Invitrogen™ R70007

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

293FT Cell Line provided as a frozen stock in 1mL of 90% complete media and 10% DMSO

Ideal for generating high-titer lentivirus using the ViraPower Lentiviral Expression System

The 293FT Cell line is a fast-growing, highly transfectable clonal isolate derived from human embryonal kidney cells transformed with the SV40 large T antigen. When a ViraPower expression vector and the ViraPower™ Lentiviral Packaging Mix are co-transfected into 293FT cells, high levels of the viral RNA and the gag/pol and rev proteins required for packaging are produced.

  • Fast growing
  • Highly transfectable
  • Produces high viral titers
  • Allows very high levels of protein
  • Presence of the SV40 large T antigen allows very high levels of protein to be expressed from vectors containing the SV40 origin

Bioproduction, Cell Culture, Constitutive Expression, Mammalian Cell Culture, Mammalian Expression, Mammalian Expression Cell Lines, Protein Expression, Proteins, Expression, Isolation and Analysis, Regulated Expression, Viral Gene Delivery, Viral Vaccine Production

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 293FT Cell Line (Store in liquid nitrogen)
Cell Line 293FT
Species Human
Media Recommendation SFM (Serum Free Medium)
No. of Cells 1 x 107
Product Line ViraPower
Quantity 1 x 107 cells

For research use only. Not intended for any therapeutic or diagnostic use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.