Learn More
Invitrogen™ Yeast Deletion Matα, Complete Set
Yeast deletion library created by a collaboration of eight North American and eight European laboratories composing the Saccharomyces Genome Deletion Project
Brand: Invitrogen™ 95401.H3
Code : NEW
Additional Details : Weight : 0.50000kg
Description
Each yeast deletion clone is supplied as a stab culture in yeast peptone dextrose (YPD) agar containing Geneticin™ (G418).
Order Info
Shipping Condition: Dry Ice
Specifications
Yeast Deletion Mat | |
Stab culture in yeast peptone dextrose (YPD) agar containing Geneticin™ (G418) | |
53 plates |
Yeast deletion library created by a collaboration of 8 North American and 8 European laboratories composing the Saccharomyces Genome Deletion Project | |
Complete Set |