missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Zero Blunt™ TOPO™ PCR Cloning Kit, with One Shot™ TOP10 Electrocomp™ E. coli
Offers the fastest and easiest method for high-efficiency cloning of blunt-end PCR products amplified with proofreading thermostable polymerases
£754.00 - £1124.00
Specifications
Bacterial or Yeast Strain | TOP10 |
---|---|
Cloning Method | Blunt TOPO™ |
Content And Storage | Box 1: • Linearized and topoisomerase I-activated pCR™Blunt II-TOPO™ vector • Salt solution • dNTPs • Control template • M13 forward and reverse primers • Sterile water Store at -5 to -30°C. All reagents are stable for 6 months when properly stored. Box 2: • One Shot™ Chemically Competent or Electrocomp™ E. coli • S.O.C. medium • Supercoiled pUC19 control plasmid Store at -68 to -85°C. |
Format | Kit |
Product Line | One Shot™ |
Includes: Linearized and topoisomerase I-activated pCR™Blunt II-TOPO™ vector; Salt solution; dNTPs; Control template; M13 forward and reverse primers; Sterile water; One Shot™ Chemically Competent or Electrocomp™ E. coli; S.O.C. medium; Supercoiled pUC19 control plasmid
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10338532
|
Invitrogen™
K286020 |
25 Reactions |
£754.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10398192
|
Invitrogen™
K286040 |
50 Reactions |
£1124.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kit includes linearized and topoisomerase I-activated pCR™-Blunt II-TOPO Vector for 5-minute benchtop ligations without ligase.
- ccdB gene for positive selection
- EcoR I sites flanking the PCR product insertion site for easy excision of inserts
- Kanamycin and Zeocin™ resistance genes for your choice of selection in E. coli
- SP6 promoter/primer site for in vitro RNA transcription and sequencing
- M13 forward and reverse primer sites for sequencing or PCR screening
- Selected Zero Blunt TOPO PCR Cloning Kits are available in combo kits combined with a PureLink™ Quick Plasmid Miniprep Kit (50 preps) for fast plasmid purification of your TOPO-cloned inserts for downstream analysis
Cloning, PCR Cloning
Specifications
TOP10 | |
Box 1: • Linearized and topoisomerase I-activated pCR™Blunt II-TOPO™ vector • Salt solution • dNTPs • Control template • M13 forward and reverse primers • Sterile water Store at -5 to -30°C. All reagents are stable for 6 months when properly stored. Box 2: • One Shot™ Chemically Competent or Electrocomp™ E. coli • S.O.C. medium • Supercoiled pUC19 control plasmid Store at -68 to -85°C. |
|
One Shot™ | |
SP6 |
Blunt TOPO™ | |
Kit | |
PCR Cloning Kit | |
Blunt DNA Cloning Vectors |
For Research Use Only. Not for use in diagnostic procedures.