missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Invivofectamine™ 3.0 Reagent
An animal origin-free lipid nanoparticle designed for high efficiency in vivo delivery of siRNA and miRNA to mouse liver cells following tail vein injection
Brand: Invitrogen™ IVF3001
1341.40 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.
This item is not returnable.
View return policy
Description
Using Invivofectamine™ 3.0 ReagentInvitrogen™ Invivofectamine™ 3.0 Reagent is ideally suited for in vivo delivery of siRNA and miRNA duplexes. Invivofectamine™ 3.0/RNA complexes are delivered using a low-volume, low-pressure method with no toxicity and no stress response in the animal. Invivofectamine™ 3.0 can also be combined with BLOCK-iT™ fluorescent controls to track the biodistribution of the siRNA or miRNA.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Animal Origin-Free | |
Synthetic siRNA, siRNA, miRNA, RNAi | |
Small Animal Models | |
10 | |
Reagent | |
siRNA, miRNA | |
Dry Ice |
Hepatocytes | |
Lipid-based Transfection, Lipid Nanoparticles, LNPs | |
Transfection | |
Invivofectamine™ | |
10 reactions | |
Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction