Learn More
Invitrogen™ Invivofectamine™ 3.0 Reagent
An animal origin-free lipid nanoparticle designed for high efficiency in vivo delivery of siRNA and miRNA to mouse liver cells following tail vein injection
Brand: Invitrogen™ IVF3005
4429.63 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.25000kg Transport : UN number : 1170 Chem class : 3 Pack group : III
Description
Using Invivofectamine™ 3.0 ReagentInvitrogen™ Invivofectamine™ 3.0 Reagent is ideally suited for in vivo delivery of siRNA and miRNA duplexes. Invivofectamine™ 3.0/RNA complexes are delivered using a low-volume, low-pressure method with no toxicity and no stress response in the animal. Invivofectamine™ 3.0 can also be combined with BLOCK-iT™ fluorescent controls to track the biodistribution of the siRNA or miRNA.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Animal Origin-Free | |
Synthetic siRNA, siRNA, miRNA, RNAi | |
Small Animal Models | |
50 Reactions | |
Reagent | |
siRNA, miRNA | |
Dry Ice |
Hepatocytes | |
Lipid-based Transfection, Lipid Nanoparticles, LNPs | |
Transfection | |
Invivofectamine™ | |
50 Reactions | |
Yes |
For Research Use Only. Not for use in diagnostic procedures.