missing translation for 'onlineSavingsMsg'
Learn More

Iodic acid, ≥99.5% (RT), Honeywell™

Product Code. 15685870 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15685870

Brand: Honeywell-Fluka 5806025G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7782-68-5
Molecular Formula ILiO3
Molecular Weight (g/mol) 181.84
MDL Number MFCD00011348
InChI Key FZAXZVHFYFGNBX-UHFFFAOYSA-M
Synonym unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min
PubChem CID 24345
ChEBI CHEBI:24857
IUPAC Name lithium(1+) iodate
SMILES [Li+].[O-][I](=O)=O

Specifications

Packaging Glass bottle
Quantity 25 g
UN Number UN3085
Formula Weight 175.91g/mol
Percent Purity ≥99.5% (RT)
Chemical Name or Material Iodic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.