missing translation for 'onlineSavingsMsg'
Learn More

Iodide Iodate concentrate, For 1L standard solution, 0.05 M I2 (0.1N), Honeywell Fluka™

for 1L standard solution, 0.05 M I2 (0.1N)

£41.38 - £267.18

Chemical Identifiers

CAS 7681-11-0, 7553-56-2, 7758-05-6, 7732-18-5
Molecular Formula IK
MDL Number MFCD00011355
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES [K+].[I-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15695200
View Documents
Honeywell-Fluka
38061-1EA
1 Pc.
£41.38
Each
Please to purchase this item. Need a web account? Register with us today!
15605210
View Documents
Honeywell Fluka
38061-6X1EA
6 x 1 Ea.
£267.18
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7681-11-0, 7553-56-2, 7758-05-6, 7732-18-5
MFCD00011355
[K+].[I-]
IK
NLKNQRATVPKPDG-UHFFFAOYSA-M

Specifications

7681-11-0, 7553-56-2, 7758-05-6, 7732-18-5
MFCD00011355
3587194
[K+].[I-]
166.00
Iodide Iodate concentrate
IK
NONH for all modes of transport
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium iodide
Ampoule
Videos
Safety and Handling

Safety and Handling

P305 + P351 + P338

SDS
Documents

Documents

Product Certifications