missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine, ACS Reagent, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
2.5 kg
6 x 500 g
Unit Size:
100g
1kg
2.5kg
500g
5g
Pack of 6
Description
Honeywell ACS solvents for general laboratory use are:
- a value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested tmeet ACS general use specifications
Specifications
Specifications
| Physical Form | Solid |
| Quantity | 1 kg |
| UN Number | 3495 |
| Beilstein | 3587194 |
| IUPAC Name | diiodine |
| Formula Weight | 253.81g/mol |
| Percent Purity | ≥99.8% |
| Grade | ACS Reagent |
| Vapor Pressure | 0.31mmHg (25°C), 1mmHg (38.7°C) |
| Density | Vapor Density: 9 (vs air) |
| Show More |
Safety and Handling
Product Identifier
- Iodine
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity Category 1
- ACUTE AQUATIC Acute 1
- H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction