missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine, Certified AR for Analysis, Fisher Chemical™
201.12 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
100 g
250 g
500 g
Specifications
Boiling Point | 185°C |
Melting Point | 113°C |
Color | Gray |
pH | 5.1 |
Physical Form | Solid |
Quantity | 500 g |
IUPAC Name | molecular iodine |
Formula Weight | 253.81g/mol |
Packaging | Amber glass bottle |
Vapor Pressure | 0.41 hPa 25 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS09_Aquatic_Toxicity.png)
Product Identifier
- Iodine
- Danger
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity Category 1
- ACUTE AQUATIC Acute 1
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P273-Avoid release to the environment.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'