missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine Concentrated Volumetric Solution Ampule, Reagecon
Fully factorized, high purity, stable product, developed and tested for titrations.
Brand: Reagecon IC025G1L
This item is not returnable.
View return policy
Description
- Use to make 1 L of 0.025 M (0.05 N) Iodine
- Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
- Accurate, stable, reproducible and fully factorized.
- Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
- Stability is proven (either opened or unopened) for the shelf life of this product.
- When the product is accurately diluted to one liter in a volumetric flask, it can then be used as an analytical volumetric solution, to a specification of ± 0.2%.
- The product must be diluted accurately to one liter, in a volumetric flask under controlled temperature conditions, for optimal performance.
- Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
- May be used for manual titrations, or automatic titrations, to determine the end points by detection of any of several properties.
Specifications
Accuracy 0.2% | |
Ampule | |
Titration |
0.025 M (0.05 N) | |
1 Ampule | |
Liquid |