missing translation for 'onlineSavingsMsg'
Learn More
Iodine monobromide, 98%, Thermo Scientific Chemicals Product Code.: 11443217

Iodine monobromide, 98%, Thermo Scientific Chemicals

Product Code. 11443217
250 g
Click to view available options
Quantity:
25 g
100 g
250 g
This item is not returnable. View return policy

Product Code. 11443217

Brand: Thermo Scientific Alfa Aesar A16158.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Iodine monobromide is used as an electrophile employed in a new synthetic approach to polyketide structural units. It is also used in iodometry and serves as a source of I+. It is a powerful iodinating agent used in organic synthesis as well as involved in the synthesis of radioiodinated fatty acids for heart imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodine monobromide is used as an electrophile employed in a new synthetic approach to polyketide structural units. It is also used in iodometry and serves as a source of I+. It is a powerful iodinating agent used in organic synthesis as well as involved in the synthesis of radioiodinated fatty acids for heart imaging.

Solubility
Soluble in water, alcohol, ether, carbon disulfide and glacial acetic acid.

Notes
Air, light and moisture sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with alcohols, phosphorus, sodium and sodiumoxides and potassium.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-33-5
Molecular Formula BrI
Molecular Weight (g/mol) 206.81
MDL Number MFCD00011353
InChI Key CBEQRNSPHCCXSH-UHFFFAOYSA-N
Synonym iodine monobromide, iodine bromide, iodobromine, iodine bromide ibr, unii-g0k622rd9n, iodinemonobromide, iodobromane, bromoiodine, jodmonobromid, hanus solutionShow MoreShow Less
PubChem CID 82238
IUPAC Name iodobromane
SMILES BrI

Specifications

Boiling Point 116°C (decomposition)
Melting Point 50°C (sublimation)
Quantity 250 g
UN Number UN3260
Beilstein 3902973
Flash Point None
Merck Index 14,5016
Solubility Information Soluble in water,alcohol,ether,carbon disulfide and glacial acetic acid.
Formula Weight 206.81
Percent Purity 98%
Odor Pungent
Sensitivity Light and moisture sensitive
Density 4.416
Chemical Name or Material Iodine monobromide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249542 Certificate TypeCertificate of Analysis Date05/03/2024 Catalog NumberA16158.30,A16158.22,A16158.14
Lot Number10246901 Certificate TypeCertificate of Analysis Date30/06/2023 Catalog NumberA16158.30,A16158,A16158.22,A16158.14
Lot Number10233159 Certificate TypeCertificate of Analysis Date06/02/2023 Catalog NumberA16158.30,A16158.22,A16158.14
Lot Number10244815 Certificate TypeCertificate of Analysis Date28/11/2022 Catalog NumberA16158.30,A16158.22,A16158.14
Lot Number10240723 Certificate TypeCertificate of Analysis Date21/05/2022 Catalog NumberA16158.30,A16158.22,A16158.14
compliance-icons compliance-icons
Product Identifier
  • Iodine monobromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Iodine bromide (IBr)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iodine monobromide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.