Learn More
Iodine monochloride, 1M solution in dichloromethane, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 370741000
49.10 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Specifications
Iodine monochloride | |
7790-99-0, 75-09-2 | |
Solution | |
ClI | |
MFCD00011354 | |
15, 5060 | |
Solubility in water: insoluble | |
ClI | |
162.35 | |
162.35 | |
Glass bottle | |
1.4200g/mL |
1, 0M solution in dichloromethane | |
Brown to Orange | |
100 mL | |
ICl | |
01,502; 08,261; 10,212; 11,268 | |
iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french | |
QZRGKCOWNLSUDK-UHFFFAOYSA-N | |
iodochlorane | |
24640 | |
0.95 to 1.05M | |
1.42 |
Safety and Handling
- Iodine monochloride, 1,0M solution in dichloromethane
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H373-May cause damage to organs.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
Precautionary Statement
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only