missing translation for 'onlineSavingsMsg'
Learn More

Iodine Monochloride, Reagent grade, ≥95%, Honeywell™

Product Code. 15651830
Click to view available options
Quantity:
5 g
100 g
250 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7790-99-0
Molecular Formula ClI
Molecular Weight (g/mol) 162.35
MDL Number MFCD00011354
InChI Key QZRGKCOWNLSUDK-UHFFFAOYSA-N
Synonym iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french
PubChem CID 24640
IUPAC Name iodochlorane
SMILES ClI

Specifications

Boiling Point >97°C
Melting Point 27°C
Color Brown to Red
Physical Form Liquid or Solid
Quantity 250 g
Linear Formula ICl
UN Number UN1792
Formula Weight 162.36g/mol
Percent Purity ≥95%
Grade Reagent
Packaging Glass Bottle
Density 3.24g/mL at 25°C
Chemical Name or Material Iodine Monochloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'