missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine Monochloride Solution, Honeywell™ Fluka™
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
Glass Bottle
Plastic Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Physical Form | Liquid |
Quantity | 100 mL |
Linear Formula | ICl |
UN Number | UN3264 |
Beilstein | 3587194 |
Formula Weight | 162.36g/mol |
Packaging | Plastic Bottle |
Density | 1.42g/mL at 25°C |
Chemical Name or Material | Iodine Monochloride Solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'