missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine Ready-to-use standardised solution 0.05mol/L Spectracer, Fisher Chemical™
Click to view available options
Quantity:
1000 mL
Packaging:
Glass Bottle
Specifications
CAS | 7732-18-5 |
Concentration | 0.05M (0.1N) |
Packaging | Glass Bottle |
Quantity | 1000 mL |
Chemical Name or Material | Iodine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'