missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine Solution, 0.05 M, Honeywell Fluka™
Click to view available options
Quantity:
100 mL
500 mL
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 500 mL |
Beilstein | 3587194 |
IUPAC Name | molecular iodine |
Formula Weight | 253.81g/mol |
Concentration or Composition (by Analyte or Components) | 0.05 M |
Density | 1.034g/mL at 25°C |
Chemical Name or Material | Iodine Solution |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- Iodine solution
- Warning
- Specific target organ toxicity after repeated exposure Category 2
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : iodine
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'