missing translation for 'onlineSavingsMsg'
Learn More

Iodine solution, Volumetric, Reag. Ph. Eur., 0.05 M I2 (0.1N), Honeywell Fluka™

Product Code. 15674790 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
2.5 L
5 L
Pack of 4
Pack of 6
This item is not returnable. View return policy

Product Code. 15674790

Brand: Honeywell-Fluka 350902.5L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Volumetric, Reag. Ph. Eur., 0.05 M I2 (0.1N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2, 7681-11-0, 7732-18-5
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
PubChem CID 807
ChEBI CHEBI:17606
SMILES II

Specifications

Quantity 2.5 L
UN Number NONH for all modes of transport
Beilstein 3587194
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
IUPAC Name diiodine
Formula Weight 253.81g/mol
Concentration or Composition (by Analyte or Components) 0.05 M
Chemical Name or Material Iodine solution
compliance-icons
Product Identifier
  • Iodine solution
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : iodine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.