missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine solution, Volumetric, Reag. Ph. Eur., 0.05 M I2 (0.1N), Honeywell Fluka™
277.03 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
6 x 1 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Quantity | 4 x 2.5 L |
UN Number | NONH for all modes of transport |
Synonym | iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode |
IUPAC Name | molecular iodine |
Formula Weight | 253.81 |
Grade | Reagent Ph. Eur. |
Chemical Name or Material | Iodine solution, volumetric, 0.05 M I2 (0.1N) |
Safety and Handling
Product Identifier
- Iodine solution
- Warning
- Specific target organ toxicity after repeated exposure Category 2
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : iodine
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'