missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine Solution 0.05M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
35.48 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Use promo code "21743" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
Description
I2, CAS Number-7553-56-2, tincture iodine, iodine sublimed, iodine solution, iodine crystals, iodine, iode, diiodine, vistarin, iodio, eranol, 1L, CHEBI:17606, Brown, PNDPGZBMCMUPRI-UHFFFAOYSA-N, molecular iodine, 0 deg.C, 11355, 253.809, Amber glass bottle, Liquid, 807, II, 0.7, 14mmHg
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Brown |
Melting Point | 0°C |
Grade | pH.Eur. |
Packaging | Amber glass bottle |
Vapor Pressure | 14mmHg |
Vapor Density | 0.7 |
pH | 5 |
Quantity | 1 L |
Physical Form | Liquid |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- Iodine solution 0.05M (0.1N)
- Warning
- Specific target organ toxicity Category 2
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P314-Get medical advice/attention if you feel unwell.
- MIXTURE LIST-Contains : Potassium iodide, Iodine
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'