missing translation for 'onlineSavingsMsg'
Learn More

Iodine, specified according to the requirements of Ph.Eur, BP, USP

Product Code. 15562323 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 15562323

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7553-56-2 | I2 | 253.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name diiodine
SMILES II

Specifications

Boiling Point 185.0°C
Melting Point 113.0°C
Quantity 250 g
Linear Formula I2
Solubility Information Solubility in water: 0.3g/L (20°C). Other solubilities: soluble in aq.sol. of hi or iodines, soluble in alcohol, carbon disulfide, chloroform,, ether, ccl4 and glycerol
Formula Weight 253.81
Percent Purity 99.8 to 100.5% (Iodometry)
Chemical Name or Material Iodine, specified according to the requirements of Ph.Eur, BP, USP

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.