missing translation for 'onlineSavingsMsg'
Learn More

Iodine spheres, ultra dry, -10 mesh, 99.998% (metals basis), Thermo Scientific Chemicals

£122.61 - £484.41

Specifications

Melting Point 183°C to 186°C (sublimation)
CAS 7553-56-2
Physical Form Solid
Molecular Formula I{2}
Percent Purity 99.998%
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11387877
View Documents
Thermo Scientific Alfa Aesar
014312.06
5 g
£122.61
5g
Please to purchase this item. Need a web account? Register with us today!
11377877
View Documents
Thermo Scientific Alfa Aesar
014312.14
25 g
£484.41
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Iodine spheres are used in OLED materials, chemicals, pharmaceuticals and electronics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

183°C to 186°C (sublimation)
Solid
99.998%
7553-56-2
I{2}
Videos
SDS
Documents

Documents

Product Certifications