Learn More
Iodoacetic acid, sodium salt, 99%, Thermo Scientific Chemicals
£89.95 - £246.23
Chemical Identifiers
CAS | 305-53-3 |
---|---|
Molecular Formula | C2H2INaO2 |
Molecular Weight (g/mol) | 207.93 |
MDL Number | MFCD00002686 |
InChI Key | AGDSCTQQXMDDCV-UHFFFAOYSA-M |
Synonym | sodium iodoacetate, iodoacetic acid sodium salt, acetic acid, iodo-, sodium salt, sodium 2-iodoacetate, sodium monoiodoacetate, iodoacetate, sodium salt, sodium iodacetate, iodoacetic acid, sodium salt, unii-07814f2qri, ccris 9232 |
PubChem CID | 5239 |
IUPAC Name | sodium;2-iodoacetate |
SMILES | C(C(=O)[O-])I.[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10235940
|
Thermo Scientific Acros
170970250 |
25 g | Glass bottle |
£89.95
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10245940
|
Thermo Scientific Acros
170971000 |
100 g | Glass bottle |
£246.23
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
305-53-3 | |
207.93 | |
AGDSCTQQXMDDCV-UHFFFAOYSA-M | |
5239 | |
C(C(=O)[O-])I.[Na+] |
C2H2INaO2 | |
MFCD00002686 | |
sodium iodoacetate, iodoacetic acid sodium salt, acetic acid, iodo-, sodium salt, sodium 2-iodoacetate, sodium monoiodoacetate, iodoacetate, sodium salt, sodium iodacetate, iodoacetic acid, sodium salt, unii-07814f2qri, ccris 9232 | |
sodium;2-iodoacetate |
Specifications
305-53-3 | |
Brown-Yellow to Yellow | |
99% | |
ICH2CO2Na | |
02, 222 | |
sodium iodoacetate, iodoacetic acid sodium salt, acetic acid, iodo-, sodium salt, sodium 2-iodoacetate, sodium monoiodoacetate, iodoacetate, sodium salt, sodium iodacetate, iodoacetic acid, sodium salt, unii-07814f2qri, ccris 9232 | |
AGDSCTQQXMDDCV-UHFFFAOYSA-M | |
sodium;2-iodoacetate | |
5239 | |
99% | |
Iodoacetic acid, sodium salt |
208.0°C to 210.0°C | |
Authentic | |
C2H2INaO2 | |
MFCD00002686 | |
13,116 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol,slightly soluble in ether | |
C(C(=O)[O-])I.[Na+] | |
207.93 | |
207.93 | |
Powder |
Safety and Handling
GHS H Statement
Toxic if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 206-165-7
missing translation for 'rtecsNumber' : AI3675000
missing translation for 'tsca' : TSCA
Recommended Storage : −20°C
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.