missing translation for 'onlineSavingsMsg'
Learn More
Iodoacetic anhydride, 97%, Thermo Scientific Chemicals Product Code.: 15179263

Iodoacetic anhydride, 97%, Thermo Scientific Chemicals

Product Code. 15179263
1 g
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15179263

Brand: Thermo Scientific Acros 307590010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54907-61-8 | C4H4I2O3 | 353.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54907-61-8
Molecular Formula C4H4I2O3
Molecular Weight (g/mol) 353.88
MDL Number MFCD00001080
InChI Key RBNSZWOCWHGHMR-UHFFFAOYSA-N
Synonym iodoacetic anhydride, di iodoacetic anhydride, acmc-20akq5, iodoacetic acid anhydride, 2-iodoacetyl 2-iodoacetate, acetic acid, iodo-, anhydride, acetic acid, 2-iodo-,1,1'-anhydride, iodoacetic anhydride, technical tShow MoreShow Less
PubChem CID 2733254
IUPAC Name (2-iodoacetyl) 2-iodoacetate
SMILES C(C(=O)OC(=O)CI)I

Specifications

Melting Point 43.0°C to 49.0°C
Color Yellow
Flash Point >110°C
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula (ICH2CO)2O
Quantity 1 g
UN Number 3261
Formula Weight 353.88
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Iodoacetic Anhydride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463200 Certificate TypeCertificate of Analysis Date01/07/2024 Catalog Number307590010,307592500
Lot NumberA0455940 Certificate TypeCertificate of Analysis Date08/11/2023 Catalog Number307590010,307592500
Lot NumberA0414392 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number307590010,307592500
Lot NumberA0406504 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number307590010,307592500
Lot NumberA0416310 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number307590010,307592500
compliance-icons compliance-icons
Product Identifier
  • Iodoacetic anhydride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Iodoacetic anhydride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iodoacetic anhydride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.