missing translation for 'onlineSavingsMsg'
Learn More

Iodoacetonitrile, 97%, Thermo Scientific Chemicals

£38.89 - £161.81

Chemical Identifiers

CAS 624-75-9
Molecular Formula C2H2IN
Molecular Weight (g/mol) 166.949
MDL Number MFCD00001886
InChI Key VODKOOOHHCAWFR-UHFFFAOYSA-N
Synonym iodoacetonitrile, acetonitrile, iodo, iodo-acetonitrile, 2-iodoethanenitrile, 2-iodo acetonitrile, 2-iodoaceto-nitrile, 2-iodanylethanenitrile, pubchem12543, acmc-1ba3n
PubChem CID 69356
IUPAC Name 2-iodoacetonitrile
SMILES C(C#N)I
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11332256
SDS missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
L03634.09
10 g
£38.89
10g
Please to purchase this item. Need a web account? Register with us today!
11342256
SDS missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
L03634.18
50 g
£161.81
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Iodoacetonitrile is used for alkylation of nucleophiles. It undergoes Reformatsky and Wittig-type reactions with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodoacetonitrile is used for alkylation of nucleophiles. It undergoes Reformatsky and Wittig-type reactions with aldehydes.

Solubility
Soluble in benzene, alcohol, acetone, ether. Slightly soluble in water. Insoluble in hexane.

Notes
Light Sensitive. Store at 4°C. Incompatible with reducing agents, light and bases. Store away from strong bases.
Specifications

Chemical Identifiers

624-75-9
166.949
VODKOOOHHCAWFR-UHFFFAOYSA-N
69356
C(C#N)I
C2H2IN
MFCD00001886
iodoacetonitrile, acetonitrile, iodo, iodo-acetonitrile, 2-iodoethanenitrile, 2-iodo acetonitrile, 2-iodoaceto-nitrile, 2-iodanylethanenitrile, pubchem12543, acmc-1ba3n
2-iodoacetonitrile

Specifications

624-75-9
70°C to 72°C (10 mmHg)
C2H2IN
1.574
UN2922
Light sensitive
Soluble in benzene,alcohol,acetone,ether. Slightly soluble in water. Insoluble in hexane.
C(C#N)I
166.949
166.95
Iodoacetonitrile
2.306
86°C (186°F)
ICH2CN
MFCD00001886
1734628
iodoacetonitrile, acetonitrile, iodo, iodo-acetonitrile, 2-iodoethanenitrile, 2-iodo acetonitrile, 2-iodoaceto-nitrile, 2-iodanylethanenitrile, pubchem12543, acmc-1ba3n
VODKOOOHHCAWFR-UHFFFAOYSA-N
2-iodoacetonitrile
69356
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H312-H332-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.

P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H227-H301+H311+H331-H314-H335

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II

missing translation for 'einecsNumber' : 210-860-0

missing translation for 'rtecsNumber' : AM0740000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

Product Identifier
  • Iodoacetonitrile

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : Iodoacetonitrile

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iodoacetonitrile, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.