Learn More
Iodobenzene, 98%, Thermo Scientific Chemicals
£29.40 - £370.00
Chemical Identifiers
CAS | 591-50-4 |
---|---|
Molecular Formula | C6H5I |
Molecular Weight (g/mol) | 204.01 |
MDL Number | MFCD00001029 |
InChI Key | SNHMUERNLJLMHN-UHFFFAOYSA-N |
Synonym | benzene, iodo, phenyl iodide, benzene iodide, iodophenyl, iodinebenzol, iodo-benzene, phenyliodide, iodo benzene, unii-9hk5l7ybbr, 1-iodobenzene |
PubChem CID | 11575 |
IUPAC Name | iodobenzene |
SMILES | IC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11495133
|
Thermo Scientific Alfa Aesar
A12788.18 |
50 g |
£29.40
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11418880
|
Thermo Scientific Alfa Aesar
A12788.22 |
100 g |
£53.30
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11485133
|
Thermo Scientific Alfa Aesar
A12788.0B |
1000 g |
£370.00
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Iodobenzene is used to prepare biphenyl and stilbene. It reacts with magnesium to form the Grignard reagent, a phenylmagnesium iodide used in organic synthesis. It finds application as a solid chlorine source by reacting with chlorine to form a complex, iodobenzene dichloride. It serves as a substrate in Sonogashira reaction which is used to form a carbon-carbon bond between a terminal alkyne and an aryl halide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIodobenzene is used to prepare biphenyl and stilbene. It reacts with magnesium to form the Grignard reagent, a phenylmagnesium iodide used in organic synthesis. It finds application as a solid chlorine source by reacting with chlorine to form a complex, iodobenzene dichloride. It serves as a substrate in Sonogashira reaction which is used to form a carbon-carbon bond between a terminal alkyne and an aryl halide.
Solubility
Miscible with ethanol, chloroform and ether. Slightly miscible with dimethyl sulfoxide. Immiscible with water.
Notes
Light sensitive. Incompatible with magnesium and strong oxidizing agents.
Chemical Identifiers
591-50-4 | |
204.01 | |
SNHMUERNLJLMHN-UHFFFAOYSA-N | |
11575 | |
IC1=CC=CC=C1 |
C6H5I | |
MFCD00001029 | |
benzene, iodo, phenyl iodide, benzene iodide, iodophenyl, iodinebenzol, iodo-benzene, phenyliodide, iodo benzene, unii-9hk5l7ybbr, 1-iodobenzene | |
iodobenzene |
Specifications
591-50-4 | |
1.826 | |
74°C (165°F) | |
C6H5I | |
MFCD00001029 | |
Light sensitive | |
benzene, iodo, phenyl iodide, benzene iodide, iodophenyl, iodinebenzol, iodo-benzene, phenyliodide, iodo benzene, unii-9hk5l7ybbr, 1-iodobenzene | |
SNHMUERNLJLMHN-UHFFFAOYSA-N | |
iodobenzene | |
11575 | |
98% |
°C to 31°C | |
188°C to 189°C | |
98% | |
1.62 | |
1446140 | |
14,5029 | |
Miscible with ethanol,chloroform and ether. Slightly miscible with dimethyl sulfoxide. Immiscible with water. | |
IC1=CC=CC=C1 | |
204.01 | |
204.01 | |
Iodobenzene |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P210-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P312-P330-P370+P378q-P501c
H227-H302+H332
missing translation for 'einecsNumber' : 209-719-6
missing translation for 'rtecsNumber' : DA3390000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only