missing translation for 'onlineSavingsMsg'
Learn More

Iodobenzene, 98%, Thermo Scientific Chemicals

Product Code. 10558850
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-50-4
Molecular Formula C6H5I
Molecular Weight (g/mol) 204.01
MDL Number MFCD00001029
InChI Key SNHMUERNLJLMHN-UHFFFAOYSA-N
Synonym benzene, iodo, phenyl iodide, benzene iodide, iodophenyl, iodinebenzol, iodo-benzene, phenyliodide, iodo benzene, unii-9hk5l7ybbr, 1-iodobenzene
PubChem CID 11575
IUPAC Name iodobenzene
SMILES IC1=CC=CC=C1

Specifications

Melting Point -29.0°C
Color Yellow
Density 1.8200g/mL
Boiling Point 188.0°C
Flash Point 74°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5I
Refractive Index 1.6180 to 1.6200
Quantity 100 mL
Beilstein ,215
Fieser 01,505
Merck Index 15,5072
Specific Gravity 1.82
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform and ether
Formula Weight 204.01
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Iodobenzene
Show More Show Less
compliance-icons
Product Identifier
  • Iodobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'