missing translation for 'onlineSavingsMsg'
Learn More

Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals

£40.60 - £549.74

Chemical Identifiers

CAS 75-03-6, 64-19-7
Molecular Formula C2H5I
Molecular Weight (g/mol) 155.97
MDL Number MFCD00001091
InChI Key HVTICUPFWKNHNG-UHFFFAOYSA-N
Synonym mercury 2+ ion acetic acid
PubChem CID 15337
ChEBI CHEBI:33211
SMILES CCI
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10023830
View Documents Promotion Details
Thermo Scientific Acros
332121000
100 g Glass bottle
£40.60
100g
Please to purchase this item. Need a web account? Register with us today!
10339420
View Documents
Thermo Scientific Acros
332125000
500 g Glass bottle
£121.61
500g
Please to purchase this item. Need a web account? Register with us today!
11964501
View Documents
Thermo Scientific Acros
332120025
2.5 kg Glass bottle
£549.74
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
Specifications

Chemical Identifiers

75-03-6, 64-19-7
155.97
HVTICUPFWKNHNG-UHFFFAOYSA-N
15337
CCI
C2H5I
MFCD00001091
mercury 2+ ion acetic acid
CHEBI:33211

Specifications

75-03-6, 64-19-7
Colorless to Yellow or Pink
69.0°C to 73.0°C
Authentic
C2H5I
MFCD00001091
15,3868
mercury 2+ ion acetic acid
HVTICUPFWKNHNG-UHFFFAOYSA-N
mercury(2+);diacetate
155.97
CHEBI:33211
155.97
Pure
Iodoethane, Stabilized
-108.0°C
1.9500g/mL
62°C
97.5% min. (GC)
1.5100 to 1.5130
01,96
1.95
Solubility in water: 4g/L (20°C),slowly decomposes in water. Other solubilities: miscible with most other organic solvents,soluble in alcohol,ether,chloroform
CCI
silver
15337
0.592 mPa.s (20°C)
98%
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing dif

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 200-833-1

missing translation for 'rtecsNumber' : KI4750000

missing translation for 'tsca' : TSCA

Recommended Storage : Product darkens on exposure to light

Product Identifier
  • Iodoethane, stabilized

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only