Learn More
Iodomethane, 99.5%, stab. with copper, Thermo Scientific Chemicals
£48.40 - £150.00
Chemical Identifiers
CAS | 74-88-4 |
---|---|
Molecular Formula | CH3I |
Molecular Weight (g/mol) | 141.94 |
MDL Number | MFCD00001073 |
InChI Key | INQOMBQAUSQDDS-UHFFFAOYSA-N |
Synonym | methyl iodide, monoiodomethane, methane, iodo, methyljodid, jod-methan, methyliodide, methyljodide, iodometano, joodmethaan, metylu jodek |
PubChem CID | 6328 |
ChEBI | CHEBI:39282 |
IUPAC Name | iodomethane |
SMILES | CI |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15451247
|
Thermo Scientific Alfa Aesar
031876.22 |
100 g |
£48.40
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15461247
|
Thermo Scientific Alfa Aesar
031876.36 |
500 g |
£150.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Iodomethane is a methylating agent used for alkylating carbon, oxygen, sulfur, nitrogen and phosphorus nucleophiles. For example, phenol reacts with methyl iodide to give anisole. In the Monsanto process, the insitu formed iodomethane reacts with carbon monoxide in the presence of a rhodium complex to give acetyl iodide, which on hydrolysis yields acetic acid. It reacts with magnesium to form the Grignard reagent, methylmagnesium iodide used in organic synthesis. It finds applications as an intermediate in the manufacture of pharmaceuticals and in phase transfer catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
74-88-4 | |
141.94 | |
INQOMBQAUSQDDS-UHFFFAOYSA-N | |
6328 | |
iodomethane |
CH3I | |
MFCD00001073 | |
methyl iodide, monoiodomethane, methane, iodo, methyljodid, jod-methan, methyliodide, methyljodide, iodometano, joodmethaan, metylu jodek | |
CHEBI:39282 | |
CI |
Specifications
74-88-4 | |
2.28 | |
99.5% | |
1.53 | |
969135 | |
methyl iodide, monoiodomethane, methane, iodo, methyljodid, jod-methan, methyliodide, methyljodide, iodometano, joodmethaan, metylu jodek | |
INQOMBQAUSQDDS-UHFFFAOYSA-N | |
iodomethane | |
6328 | |
141.94 | |
Iodomethane |
-67°C | |
41°C to 43°C | |
CH3I | |
MFCD00001073 | |
Light sensitive | |
Miscible with water. | |
CI | |
141.94 | |
CHEBI:39282 | |
Liquid |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P332+P313-P362-P501c
H301+H331-H312-H315-H319-H335-H351
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: I
missing translation for 'einecsNumber' : 200-819-5
missing translation for 'rtecsNumber' : PA9450000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only