missing translation for 'onlineSavingsMsg'
Learn More

Iodomethane, 99%, stabilized, Thermo Scientific Chemicals

Product Code. 15530604
Click to view available options
Quantity:
100 g
500 g
2.5 kg

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-88-4, 7440-50-8
Molecular Formula CH3I
Molecular Weight (g/mol) 141.94
MDL Number MFCD00001073
InChI Key INQOMBQAUSQDDS-UHFFFAOYSA-N
Synonym methyl iodide, monoiodomethane, methane, iodo, methyljodid, jod-methan, methyliodide, methyljodide, iodometano, joodmethaan, metylu jodek
PubChem CID 6328
ChEBI CHEBI:39282
IUPAC Name iodomethane
SMILES CI

Specifications

Melting Point -66.0°C
Density 2.2800g/mL
Boiling Point 42.5°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Linear Formula CH3I
Refractive Index 1.5290 to 1.5310
Quantity 2.5 kg
Specific Gravity 2.28
Solubility Information Solubility in water: 14g/L (20°C). Other solubilities: miscible with alcohol and ether, soluble in benzene, acetone, methylenechloride, n,n-dimethylformamide
Viscosity 0.5 mPa.s (20°C)
Formula Weight 141.94
Percent Purity 99%
Chemical Name or Material Iodomethane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iodomethane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H351-Suspected of causing cancer.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'