Learn More
Iodomethane, 99%, stabilized, Thermo Scientific Chemicals
£41.20 - £453.00
Chemical Identifiers
CAS | 74-88-4, 7440-50-8 |
---|---|
Molecular Formula | CH3I |
Molecular Weight (g/mol) | 141.94 |
MDL Number | MFCD00001073 |
InChI Key | INQOMBQAUSQDDS-UHFFFAOYSA-N |
Synonym | methyl iodide, monoiodomethane, methane, iodo, methyljodid, jod-methan, methyliodide, methyljodide, iodometano, joodmethaan, metylu jodek |
PubChem CID | 6328 |
ChEBI | CHEBI:39282 |
IUPAC Name | iodomethane |
SMILES | CI |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15582790
|
Thermo Scientific Acros
459011000 |
100 g |
£41.20
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15550604
|
Thermo Scientific Acros
459015000 |
500 g |
£138.69
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15530604
|
Thermo Scientific Acros
459010025 |
2.5 kg |
£453.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
74-88-4, 7440-50-8 | |
141.94 | |
INQOMBQAUSQDDS-UHFFFAOYSA-N | |
6328 | |
iodomethane |
CH3I | |
MFCD00001073 | |
methyl iodide, monoiodomethane, methane, iodo, methyljodid, jod-methan, methyliodide, methyljodide, iodometano, joodmethaan, metylu jodek | |
CHEBI:39282 | |
CI |
Specifications
74-88-4, 7440-50-8 | |
2.2800g/mL | |
Authentic | |
CH3I | |
1.5290 to 1.5310 | |
2.28 | |
Solubility in water: 14g/L (20°C). Other solubilities: miscible with alcohol and ether, soluble in benzene, acetone, methylenechloride, n,n-dimethylformamide | |
CI | |
141.94 | |
CHEBI:39282 | |
141.94 | |
Iodomethane |
-66.0°C | |
42.5°C | |
98.5% min. (GC) | |
CH3I | |
MFCD00001073 | |
methyl iodide, monoiodomethane, methane, iodo, methyljodid, jod-methan, methyliodide, methyljodide, iodometano, joodmethaan, metylu jodek | |
INQOMBQAUSQDDS-UHFFFAOYSA-N | |
iodomethane | |
6328 | |
0.5 mPa.s (20°C) | |
99% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Toxic if inhaled.
May cause respiratory irritation.
Suspected of causing cancer.
GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of s
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-819-5
- Iodomethane
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H351-Suspected of causing cancer.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H301-Toxic if swallowed.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
RUO – Research Use Only