missing translation for 'onlineSavingsMsg'
Learn More

Iodomethane-d3, for NMR, 99+ atom % D, Thermo Scientific Chemicals

Product Code. 10780871
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-50-9
Molecular Formula CH3I
Molecular Weight (g/mol) 144.96
MDL Number MFCD00001074
InChI Key INQOMBQAUSQDDS-FIBGUPNXSA-N
Synonym iodomethane-d3, iodo 2h3 methane, trideuterio iodo methane, iodo 2 h? methane, methyl-d3 iodide, trideuteromethyl iodide, iodomethane-12c,d3, methyl-12c,d3 iodide, iodoethane-d5 ethyliodid-d5, methyl iodide d3 stored over copper
PubChem CID 2723978
IUPAC Name trideuterio(iodo)methane
SMILES [2H]C([2H])([2H])I

Specifications

Physical Form Liquid
Color Colorless
Density 2.3290g/mL
Deuterium (D) 99.0% min.
Packaging Glass Bottle
Refractive Index 1.5262
Water 0.05% max. (Coulometric)
Quantity 5 g
Boiling Point 42.0°C
Linear Formula CD3I
Formula Weight 144.96
Grade NMR
Chemical Name or Material Iodomethane-d3, 99+ atom % D
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iodomethane-d3
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H330-Fatal if inhaled.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'