missing translation for 'onlineSavingsMsg'
Learn More

(Iodomethyl)trimethylammonium iodide, 98+%, Thermo Scientific Chemicals

Product Code. 11448347
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39741-91-8
Molecular Formula C4H11I2N
Molecular Weight (g/mol) 326.948
MDL Number MFCD00031647
InChI Key IFVIWWQMNYZYFQ-UHFFFAOYSA-M
Synonym iodomethyltrimethylammonium iodide, iodomethyl trimethylammonium iodide, ammonium, iodomethyltrimethyl-, iodide, iodomethyl trimethylazanium iodide, 1-iodo-n,n,n-trimethylmethanaminium iodide, acmc-20akpy, iodomethyltrimethylammoniumiodide, iodomethyl trimethylamine, iodide, iodomethyl trimethyl azanium iodide, iodomethyl trimethylammonium iodide, 98+%
PubChem CID 38328
IUPAC Name iodomethyl(trimethyl)azanium;iodide
SMILES C[N+](C)(C)CI.[I-]

Specifications

Melting Point ∼190°C (decomposition)
Quantity 5 g
Beilstein 3912137
Sensitivity Hygroscopic; Light sensitive
Formula Weight 326.94
Percent Purity ≥98%
Chemical Name or Material (Iodomethyl)trimethylammonium iodide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.