missing translation for 'onlineSavingsMsg'
Learn More
Iodotrimethylsilane, 95-97%, stabilized, Thermo Scientific Chemicals Product Code.: 13468428

Iodotrimethylsilane, 95-97%, stabilized, Thermo Scientific Chemicals

Product Code. 13468428
250 mL, Glass Bottle
Click to view available options
Quantity:
5 mL
25 mL
100 mL
250 mL
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 13468428

Brand: Thermo Scientific Acros 187492500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16029-98-4
Molecular Formula C3H9ISi
Molecular Weight (g/mol) 200.1
MDL Number MFCD00001028
InChI Key CSRZQMIRAZTJOY-UHFFFAOYSA-N
Synonym trimethylsilyl iodide, trimethyliodosilane, silane, iodotrimethyl, iodo trimethyl silane, trimethylsilyliodide, tms-iodide, iodotrimethysilane, iodotrimetylsilane, tms iodide, iodo trimethylsilaneShow MoreShow Less
PubChem CID 85247
IUPAC Name iodo(trimethyl)silane
SMILES C[Si](C)(C)I

Specifications

Color Colorless to Red
Density 1.4000g/mL
Boiling Point 106.0°C
Flash Point −31°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (Ex I) (Argentometry)
Packaging Glass Bottle
Linear Formula (CH3)3SiI
Refractive Index 1.4700 to 1.4800
Quantity 250 mL
UN Number 2924
Fieser 08,261; 09,251; 10,216; 11,271; 12,259; 13,329; 16,188; 06,81
Specific Gravity 1.4
Solubility Information Solubility in water: reacts
Stabilizer copper
Formula Weight 200.1
Percent Purity 95 to 97%
Physical Form Liquid
Chemical Name or Material Iodotrimethylsilane, 95-97%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462970 Certificate TypeCertificate of Analysis Date23/05/2024 Catalog Number187492500,187490250,187490050,187491000
Lot NumberA0458947 Certificate TypeCertificate of Analysis Date08/01/2024 Catalog Number187492500,187490250,187490050,187491000
Lot NumberA0444917 Certificate TypeCertificate of Analysis Date01/09/2023 Catalog Number187492500,187490250,187490050,187491000
Lot NumberA0454659 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number187492500,187490250,187490050,187491000
Lot NumberA0394053 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number187492500,187490250,187490050,187491000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iodotrimethylsilane, 95-97%, stabilized, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.