Learn More
Iodotrimethylsilane, 97%, stab. with copper, Thermo Scientific Chemicals
£23.22 - £467.33
Chemical Identifiers
CAS | 16029-98-4 |
---|---|
Molecular Formula | C3H9ISi |
Molecular Weight (g/mol) | 200.094 |
MDL Number | MFCD00001028 |
InChI Key | CSRZQMIRAZTJOY-UHFFFAOYSA-N |
Synonym | trimethylsilyl iodide, trimethyliodosilane, silane, iodotrimethyl, iodo trimethyl silane, trimethylsilyliodide, tms-iodide, iodotrimethysilane, iodotrimetylsilane, tms iodide, iodo trimethylsilane |
PubChem CID | 85247 |
IUPAC Name | iodo(trimethyl)silane |
SMILES | C[Si](C)(C)I |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11488870
|
Thermo Scientific Alfa Aesar
A12902.06 |
5 g |
£23.22
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11498870
|
Thermo Scientific Alfa Aesar
A12902.14 |
25 g |
£73.47
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11408880
|
Thermo Scientific Alfa Aesar
A12902.22 |
100 g |
£212.06
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11485353
|
Thermo Scientific Alfa Aesar
A12902.30 |
250 g |
£467.33
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Iodotrimethylsilane is used for the introduction of trimethylsilyl group in organic synthesis. It is also useful for gas chromatography analysis by converting alcohol into a silyl ether derivative, thereby making it more volatile than the original molecule.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIodotrimethylsilane is used for the introduction of trimethylsilyl group in organic synthesis. It is also useful for gas chromatography analysis by converting alcohol into a silyl ether derivative, thereby making it more volatile than the original molecule.
Solubility
Miscible with water.
Notes
Moisture and light Sensitive. Incompatible withstrong acids, strong bases, strong oxidizing agents and reacts violently with water.
Chemical Identifiers
16029-98-4 | |
200.094 | |
CSRZQMIRAZTJOY-UHFFFAOYSA-N | |
85247 | |
C[Si](C)(C)I |
C3H9ISi | |
MFCD00001028 | |
trimethylsilyl iodide, trimethyliodosilane, silane, iodotrimethyl, iodo trimethyl silane, trimethylsilyliodide, tms-iodide, iodotrimethysilane, iodotrimetylsilane, tms iodide, iodo trimethylsilane | |
iodo(trimethyl)silane |
Specifications
105°C to 106°C | |
C3H9ISi | |
MFCD00001028 | |
1731136 | |
trimethylsilyl iodide, trimethyliodosilane, silane, iodotrimethyl, iodo trimethyl silane, trimethylsilyliodide, tms-iodide, iodotrimethysilane, iodotrimetylsilane, tms iodide, iodo trimethylsilane | |
CSRZQMIRAZTJOY-UHFFFAOYSA-N | |
iodo(trimethyl)silane | |
85247 | |
97% | |
Light and moisture sensitive | |
Iodotrimethylsilane, stabilized with copper |
16029-98-4 | |
(CH3)3SiI | |
UN2924 | |
−2°C (28°F) | |
Miscible with water. | |
C[Si](C)(C)I | |
200.094 | |
200.09 | |
1.476 | |
1.44 |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 240-171-0
missing translation for 'tsca' : Yes
Recommended Storage : Store at -20°C; Store under Nitrogen
- Iodotrimethylsilane
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- EUH014-Reacts violently with water.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
- MIXTURE LIST-Contains : Iodotrimethylsilane
RUO – Research Use Only