missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ion Chromatography Fluoride Standard, 100μg/mL F-, SPEX CertiPrep™
£59.40 - £355.00
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11435938
|
SPEX Certiprep
MN-6-500 |
125 mL | Triple-leached LDPE bottle |
£355.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10224482
|
SPEX Certiprep
AS-F9-1Y |
125 mL | Triple-leached LDPE bottle |
£59.40
125mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15698225
|
SPEX Certiprep
AS-F9-1X |
500 mL | Poly Bottle |
£131.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
F- | 100μg/mL |
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Chemical Identifiers
7732-18-5 |
Specifications
7732-18-5 | |
0.01, 99.99 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
100μg/mL F− | |
Fluoride Standard |
0.01, 99.99 | |
With Certificate of Analysis. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
H2O |
Safety and Handling
missing translation for 'dotInformation' : Nonhazardous
Recommended Storage : Ambient
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Trace Metals III
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : nitric acid